Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

586366

Sigma-Aldrich

Carumonam disodium salt

97%

Synonym(s):

(+)-(Z)-(((1-(2-Amino-4-thiazolyl)-2-(((2S,3S)-2-(carbamoyl-oxymethyl)-4-oxo-1-sulfonate-3-azetidinyl)amino)-2-oxoethylidene)amino)oxy)acetate disodium salt, (+)-[[2Z-[2-[[(2S,3S)-2-[[(Aminocarbonyl)oxy]methyl]-4-oxo-1-sulfo-3-azetidinyl]amino]-1-(2-amino-4-thiazolyl)-2-oxoethylidene]amino]oxy]acetic acid disodium salt, [2S-[2α,3α(Z)]]-[[[2-[[2-[[(Aminocarbonyl)oxy]methyl]-4-oxo-1-sulfo-3-azetidinyl]amino]-1-(2-amino-4-thiozolyl)-2-oxoethylidene]amino]oxy]acetic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N6Na2O10S2
CAS Number:
Molecular Weight:
510.37
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

97%

optical activity

[α]20/D +18°, c = 1% in H2O

mp

250 °C (dec.) (lit.)

SMILES string

[Na+].[Na+].NC(=O)OC[C@@H]1[C@H](NC(=O)\C(=N/OCC([O-])=O)c2csc(N)n2)C(=O)N1S([O-])(=O)=O

InChI

1S/C12H14N6O10S2.2Na/c13-11-15-4(3-29-11)7(17-28-2-6(19)20)9(21)16-8-5(1-27-12(14)23)18(10(8)22)30(24,25)26;;/h3,5,8H,1-2H2,(H2,13,15)(H2,14,23)(H,16,21)(H,19,20)(H,24,25,26);;/q;2*+1/p-2/b17-7-;;/t5-,8+;;/m1../s1

InChI key

BGGXRVPCJUKHTQ-AHCAJXDVSA-L

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service