Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

586250

Sigma-Aldrich

(2R,3R)-Diethyl 2,3-epoxysuccinate

96%

Synonym(s):

Diethyl (2R,3R)-2,3-oxiranedicarboxylate, Diethyl-(2R,3R)-(-)-2,3-epoxysuccinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O5
CAS Number:
Molecular Weight:
188.18
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

96%

optical activity

[α]20/D −110°, c = 2% in diethyl ether

refractive index

n20/D 1.4380 (lit.)

bp

234-235 °C (lit.)

density

1.144 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)[C@@H]1O[C@H]1C(=O)OCC

InChI

1S/C8H12O5/c1-3-11-7(9)5-6(13-5)8(10)12-4-2/h5-6H,3-4H2,1-2H3/t5-,6-/m1/s1

InChI key

LDFQMMUIJQDSAB-PHDIDXHHSA-N

Application

(2R,3R)-Diethyl 2,3-epoxysuccinate may be used in the preparation of ethyl (2R,3S)-4-hydroxy-2,3-epoxybutyrate via reduction using sodium borohydride

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel synthesis of the monobactam antibiotic carumonam.
The Journal of Organic Chemistry, 53(23), 5507-5512 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service