578681
Grubbs Catalyst® C801
Synonym(s):
Bis(tricyclohexylphosphine)isopentenylidene ruthenium dichloride, Grubbs Catalyst 1st Generation 801
About This Item
Recommended Products
reaction suitability
reagent type: catalyst
core: ruthenium
mp
160-165 °C (lit.)
storage temp.
2-8°C
SMILES string
Cl[Ru]Cl.[CH][CH][C](C)C.C1CCC(CC1)P(C2CCCCC2)C3CCCCC3.C4CCC(CC4)P(C5CCCCC5)C6CCCCC6
InChI
1S/2C18H33P.C5H8.2ClH.Ru/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-4-5(2)3;;;/h2*16-18H,1-15H2;1,4H,2-3H3;2*1H;/q;;;;;+2/p-2
InChI key
INCDKKABOVOFIV-UHFFFAOYSA-L
Application
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Flam. Sol. 1
Storage Class
4.1B - Flammable solid hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service