Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

575976

Sigma-Aldrich

Tris(ethylenediamine)nickel(II) chloride hydrate

99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H24Cl2N6Ni · xH2O
CAS Number:
Molecular Weight:
309.89 (anhydrous basis)
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

99.999% trace metals basis

mp

170 °C (dec.) (lit.)

SMILES string

[H]O[H].Cl[Ni]Cl.NCCN.NCCN.NCCN

pictograms

Health hazardExclamation mark

signalword

Warning

hcodes

pcodes

Hazard Classifications

Carc. 2 - Skin Sens. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service