Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

575925

Sigma-Aldrich

Tetramethylammonium dichloroiodate(I)

99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(Cl2I)
CAS Number:
Molecular Weight:
271.96
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

assay

99.999% trace metals basis

form

powder and chunks

mp

225-229 °C (lit.)

SMILES string

Cl[I-]Cl.C[N+](C)(C)C

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service