Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

574333

Sigma-Aldrich

Iodobis(indenyl)vanadium(III)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14IV
CAS Number:
Molecular Weight:
408.15
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

reaction suitability

core: vanadium
reagent type: catalyst

mp

>300 °C (lit.)

SMILES string

I[V](C1C=Cc2ccccc12)C3C=Cc4ccccc34

InChI

1S/2C9H7.HI.V/c2*1-2-5-9-7-3-6-8(9)4-1;;/h2*1-7H;1H;/q;;;+1/p-1

InChI key

PAIIOIYTKDHMHA-UHFFFAOYSA-M

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service