All Photos(1)
About This Item
Linear Formula:
SrC2O4
CAS Number:
Molecular Weight:
175.64
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Recommended Products
assay
99.999% trace metals basis
form
powder
SMILES string
O=C1O[Sr]OC1=O
InChI
1S/C2H2O4.Sr/c3-1(4)2(5)6;/h(H,3,4)(H,5,6);/q;+2/p-2
InChI key
KQAGKTURZUKUCH-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service