Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

572071

Sigma-Aldrich

9,9-Di(2-ethylhexyl)fluorene-2,7-diboronic acid bis(1,3-propanediol) ester solution

Synonym(s):

9,9-Di(2′-ethylhexyl)fluorene-2,7-bis(trimethylene borate) solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H52B2O4
CAS Number:
Molecular Weight:
558.41
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

concentration

0.5 M in toluene

refractive index

n20/D 1.5160

bp

117-118 °C

density

0.906 g/mL at 25 °C

SMILES string

CCCCC(CC)CC1(CC(CC)CCCC)c2cc(ccc2-c3ccc(cc13)B4OCCCO4)B5OCCCO5

InChI

1S/C35H52B2O4/c1-5-9-13-27(7-3)25-35(26-28(8-4)14-10-6-2)33-23-29(36-38-19-11-20-39-36)15-17-31(33)32-18-16-30(24-34(32)35)37-40-21-12-22-41-37/h15-18,23-24,27-28H,5-14,19-22,25-26H2,1-4H3

InChI key

LGBZRFAFRYBGTM-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

55.4 °F - closed cup

flash_point_c

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service