Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

568988

Sigma-Aldrich

2-Bromo-4,6-dimethylphenyl isocyanate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H2BrNCO
CAS Number:
Molecular Weight:
226.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

96%

refractive index

n20/D 1.5750 (lit.)

bp

261-262 °C (lit.)

SMILES string

Cc1cc(C)c(N=C=O)c(Br)c1

InChI

1S/C9H8BrNO/c1-6-3-7(2)9(11-5-12)8(10)4-6/h3-4H,1-2H3

InChI key

WOURXNWXJAEYAZ-UHFFFAOYSA-N

Caution

Refrigerate

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service