Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

568678

Sigma-Aldrich

2-Mercaptoethanol

99%

Synonym(s):

β-Mercaptoethanol, 2-Hydroxyethylmercaptan, BME, Thioethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2OH
CAS Number:
Molecular Weight:
78.13
Beilstein/REAXYS Number:
773648
EC Number:
MDL number:
UNSPSC Code:
12190000

vapor density

2.69 (vs air)

vapor pressure

1 mmHg ( 20 °C)

assay

99%

expl. lim.

18 %

concentration

14.3 M (pure liquid)

refractive index

n20/D 1.500 (lit.)

bp

157 °C (lit.)

density

1.114 g/mL at 25 °C (lit.)

SMILES string

OCCS

InChI

1S/C2H6OS/c3-1-2-4/h3-4H,1-2H2

InChI key

DGVVWUTYPXICAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

BME is suitable for reducing protein disulfide bonds prior to polyacrylamide gel electrophoresis and is usually included in a sample buffer for SDS-PAGE at a concentration of 5%. Cleaving intermolecular (between subunits) disulfide bonds allows the subunits of a protein to separate independently on SDS-PAGE. Cleaving intramolecular (within subunit) disulfide bonds allows the subunits to become completely denatured so that each peptide migrates according to its chain length with no influence due to secondary structure.

Legal Information

Product of Arkema Inc.

replaced by

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1A - STOT RE 2 Oral

target_organs

Liver,Heart

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

165.2 °F - closed cup

flash_point_c

74 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leon Tribolet et al.
The Journal of infectious diseases, 211(3), 416-425 (2014-08-21)
Na-ASP-2 is an efficacious hookworm vaccine antigen. However, despite elucidation of its crystal structure and studies addressing its immunobiology, the function of Na-ASP-2 has remained elusive. We probed a 9000-protein human proteome microarray with Na-ASP-2 and showed binding to CD79A
Katharina Rothe et al.
Blood, 123(23), 3622-3634 (2014-04-24)
Previous studies demonstrated that imatinib mesylate (IM) induces autophagy in chronic myeloid leukemia (CML) and that this process is critical to cell survival upon therapy. However, it is not known if the autophagic process differs at basal levels between CML
D Zhu et al.
Leukemia, 29(7), 1587-1599 (2015-02-14)
The pathogenesis of Chlamydophila psittaci-negative ocular adnexal extranodal marginal zone lymphomas (OAEMZLs) is poorly understood. OAEMZLs are monoclonal tumors expressing a biased repertoire of mutated surface immunoglobulins. Antigenic activation of the B-cell receptor (BCR) may have a role in the
E M Michalak et al.
Cell death and differentiation, 15(6), 1019-1029 (2008-02-09)
The ability of p53 to induce apoptosis in cells with damaged DNA is thought to contribute greatly to its tumour suppressor function. P53 has been proposed to induce apoptosis via numerous transcriptional targets or even by direct cytoplasmic action. Two
Philipp Müller et al.
Science translational medicine, 7(315), 315ra188-315ra188 (2015-11-27)
Targeted drug delivery with antibody-drug conjugates such as the HER2-directed ado-trastuzumab emtansine (T-DM1) has emerged as a powerful strategy for cancer therapy. We show that T-DM1 is particularly effective in eliciting antitumor immunity in patients with early breast cancer (WSG-ADAPT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service