Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

567485

Sigma-Aldrich

3,5-Diformylphenylboronic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(CHO)2B(OH)2
CAS Number:
Molecular Weight:
177.95
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

≥95%

form

solid

mp

255-259 °C (Dec.) (lit.)

functional group

aldehyde

SMILES string

[H]C(=O)c1cc(cc(c1)C([H])=O)B(O)O

InChI

1S/C8H7BO4/c10-4-6-1-7(5-11)3-8(2-6)9(12)13/h1-5,12-13H

InChI key

RIFBPKDFVPXQQH-UHFFFAOYSA-N

Application

May contain varying amounts of anhydrides
Reactant involved in:
  • Studies of surface-tension of enzyme inhibitors
  • Synthesis of bacteriochlorins
  • Studies of the activation parameters for recomination reactions of intramolecular redical pairs
  • Reactivity studies with diethanolamine
  • Regioselective 15-bromination and functionalization of bacteriochlorins
  • Synthesis of multicomponent assemblies of boron-based dendritic nanostructures

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service