Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

560103

Sigma-Aldrich

Dichloromethane-d2

99.9 atom % D

Synonym(s):

Dideuteromethylenechloride, Methylene chloride-d2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD2Cl2
CAS Number:
Molecular Weight:
86.94
Beilstein/REAXYS Number:
1733318
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

99.9 atom % D

Quality Level

assay

≥99.00% (GC)

packaging

pk of 10 × 1 mL

refractive index

n20/D 1.422 (lit.)

bp

40 °C (lit.)

density

1.362 g/mL at 25 °C (lit.)

mass shift

M+2

SMILES string

[2H]C([2H])(Cl)Cl

InChI

1S/CH2Cl2/c2-1-3/h1H2/i1D2

InChI key

YMWUJEATGCHHMB-DICFDUPASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dichloromethane-d2 (CD2Cl2) is a deuterated NMR solvent useful in NMR-based research and analyses. Raman spectral analysis of CD2Cl2 in the gas phase has been reported.[1]

Application

Dichloromethane-d2 may be used to analyze the three-dimensional structure of (9S,11S)-11-amino-9-deoxo-11,12-deoxy-9,12-epoxyerythromycin by proton NMR derived constrained molecular mechanics.[2]

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Absolute Raman intensities, force constants, and electro-optical parameters of CH2Cl2, CD2Cl2 and CHDCl2.
Escribano R, et al.
Molecular Physics, 37(2), 361-377 (1979)
Determination of the three-dimensional, solution-phase structure of the macrolide antibiotic oxolide in CD2Cl2 and D2O from NMR constraints.
Steinmetz WE, et al.
Magnetic Resonance in Chemistry, 43(1), 16-20 (2005)
Ting Li et al.
Environmental science and pollution research international, 22(15), 11792-11800 (2015-04-11)
We surveyed the occurrence of nine N-nitrosamine species in ten bottled drinking waters from supermarket and other water samples including raw waters, finished waters, and distribution system waters from nine municipal drinking water treatment plants in eight cities of Jiangsu
Alejandro G Lichtscheidl et al.
Dalton transactions (Cambridge, England : 2003), 44(36), 16156-16163 (2015-08-22)
The synthesis and full characterization, including Nuclear Magnetic Resonance (NMR) data ((1)H, (13)C{(1)H} and (119)Sn{(1)H}), for a series of Me3SnX (X = O-2,6-(t)Bu2C6H3 (), (Me3Sn)N(2,6-(i)Pr2C6H3) (), NH-2,4,6-(t)Bu3C6H2 (), N(SiMe3)2 (), NEt2, C5Me5 (), Cl, Br, I, and SnMe3) compounds in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service