Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

559075

Sigma-Aldrich

Ethyl (2-iodobenzoyl)acetate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4COCH2CO2C2H5
CAS Number:
Molecular Weight:
318.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

95%

refractive index

n20/D 1.5865 (lit.)

bp

142-144 °C/0.4 mmHg (lit.)

density

1.609 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(=O)c1ccccc1I

InChI

1S/C11H11IO3/c1-2-15-11(14)7-10(13)8-5-3-4-6-9(8)12/h3-6H,2,7H2,1H3

InChI key

RMXYWHBYIQLPGY-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service