Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

558087

Sigma-Aldrich

Acetophenone tosylhydrazone

97%

Synonym(s):

p-Toluenesulfonic acid (α-methylbenzylidene)hydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(CH3)=NNHSO2C6H4CH3
CAS Number:
Molecular Weight:
288.36
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

solid

mp

144-149 °C (lit.)

SMILES string

Cc1ccc(cc1)S(=O)(=O)N\N=C(/C)c2ccccc2

InChI

1S/C15H16N2O2S/c1-12-8-10-15(11-9-12)20(18,19)17-16-13(2)14-6-4-3-5-7-14/h3-11,17H,1-2H3/b16-13+

InChI key

MIXFDFCKBMCLGN-DTQAZKPQSA-N

General description

Acetophenone tosylhydrazone is formed as one of the reaction products from the reduction of phenacyl in the presence of tosyl hydrazide methanol solution.[1]

Application

Acetophenone tosylhydrazone may be used to synthesize ethylbenzene, via reduction with bis(benzoyloxy)borane.[2]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 1, 2, 3-Triazoles.
Dolly BB and Barba F.
Heterocycles, 63, 1175-1180 (2004)
A mild and convenient conversion of ketones to the corresponding methylene derivatives via reduction of tosylhydrazones by bis (benzoyloxy) borane.
Kabalka GW and Summers ST.
The Journal of Organic Chemistry, 46(6), 1217-1218 (1981)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service