55708
4-Hydroxymethyl-3-methoxyphenoxyacetic acid
≥97.0%
Synonym(s):
HMPA
About This Item
assay
≥97.0%
reaction suitability
reagent type: cross-linking reagent
mp
~95 °C (dec.)
SMILES string
COc1cc(OCC(O)=O)ccc1CO
InChI
1S/C10H12O5/c1-14-9-4-8(15-6-10(12)13)3-2-7(9)5-11/h2-4,11H,5-6H2,1H3,(H,12,13)
InChI key
ZKDXHLFLKKWCBY-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service