Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

555258

Sigma-Aldrich

ScavengePore benzyl isocyanate

macroporous, 40-70 mesh, extent of labeling: 0.5-2.0 mmol/g loading

Synonym(s):

Isocyanate, polymer-bound, MP-NCO

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

product line

ScavengePore

form

macroporous

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: amine reactive

extent of labeling

0.5-2.0 mmol/g loading

particle size

40-70 mesh

storage temp.

2-8°C

SMILES string

C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.O=C=NCc3ccccc3

Legal Information

ScavengePore is a trademark of Rapp Polymere GmbH

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service