Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

554022

Sigma-Aldrich

2,5-Dimethyl-1,3,4-thiadiazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2S
CAS Number:
Molecular Weight:
114.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

mp

35-39 °C (lit.)

SMILES string

Cc1nnc(C)s1

InChI

1S/C4H6N2S/c1-3-5-6-4(2)7-3/h1-2H3

InChI key

JXQGICFGPUAVLJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service