Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

553476

Sigma-Aldrich

Bisphenol C

98%

Synonym(s):

Bis(4-hydroxyphenyl)-2,2-dichloroethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C=C(C6H4OH)2
CAS Number:
Molecular Weight:
281.13
Beilstein/REAXYS Number:
2052978
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

assay

98%

mp

213-217 °C (lit.)

SMILES string

Oc1ccc(cc1)\C(=C(/Cl)Cl)c2ccc(O)cc2

InChI

1S/C14H10Cl2O2/c15-14(16)13(9-1-5-11(17)6-2-9)10-3-7-12(18)8-4-10/h1-8,17-18H

InChI key

OWEYKIWAZBBXJK-UHFFFAOYSA-N

Application

Used in the preparation of flame-resistant polycarbonates; Modifier to form thermosets with improved flame resistant.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justin M Conley et al.
Toxicological sciences : an official journal of the Society of Toxicology, 153(2), 382-395 (2016-07-31)
In vitro estrogen receptor assays are valuable tools for identifying environmental samples and chemicals that display estrogenic activity. However, in vitro potency cannot necessarily be extrapolated to estimates of in vivo potency because in vitro assays are currently unable to
Vanessa Delfosse et al.
Environmental health perspectives, 122(12), 1306-1313 (2014-09-27)
Individuals are exposed daily to environmental pollutants that may act as endocrine-disrupting chemicals (EDCs), causing a range of developmental, reproductive, metabolic, or neoplastic diseases. With their mostly hydrophobic pocket that serves as a docking site for endogenous and exogenous ligands
Caroline Pinto et al.
Toxicology and applied pharmacology, 380, 114709-114709 (2019-08-16)
The high volume production compound bisphenol A (BPA) is of environmental concern largely because of its estrogenic activity. Consequently, BPA analogues have been synthesized to be considered as replacement molecules for BPA. These analogues need to be thoroughly evaluated for
Xiaohui Liu et al.
Toxicology and applied pharmacology, 377, 114610-114610 (2019-06-14)
An endocrine-disrupting chemical Bisphenol A (BPA) binds specifically to a nuclear receptor (NR) named ERRγ. Although the importance of receptor-binding evaluation for human NRs is often stressed, the binding characteristics of so-called next-generation (NextGen) bisphenol compounds are still poorly understood.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service