552526
(S)-(−)-α,α-Diphenyl-2-pyrrolidinemethanol
98%
Synonym(s):
(S)-(−)-2-(Diphenylhydroxymethyl)pyrrolidine, α,α-Diphenyl-L-prolinol
About This Item
assay
98%
optical activity
[α]20/D −67°, c = 3 in chloroform
mp
77-80 °C (lit.)
SMILES string
OC([C@@H]1CCCN1)(c2ccccc2)c3ccccc3
InChI
1S/C17H19NO/c19-17(16-12-7-13-18-16,14-8-3-1-4-9-14)15-10-5-2-6-11-15/h1-6,8-11,16,18-19H,7,12-13H2/t16-/m0/s1
InChI key
OGCGXUGBDJGFFY-INIZCTEOSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service