549290
2,2′-Diaminophenylsulfide
97%
Synonym(s):
2,2′-Thiodianiline
About This Item
assay
97%
mp
85-88 °C (lit.)
SMILES string
Nc1ccccc1Sc2ccccc2N
InChI
1S/C12H12N2S/c13-9-5-1-3-7-11(9)15-12-8-4-2-6-10(12)14/h1-8H,13-14H2
Inchi Key
WRRQKFXVKRQPDB-UHFFFAOYSA-N
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service