Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

548111

Sigma-Aldrich

Heptafluorobenzyl iodide

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7F7I
CAS Number:
Molecular Weight:
343.97
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

refractive index

n20/D 1.47 (lit.)

bp

153 °C (lit.)

density

2.185 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Fc1c(F)c(F)c(c(F)c1F)C(F)(F)I

InChI

1S/C7F7I/c8-2-1(7(13,14)15)3(9)5(11)6(12)4(2)10

InChI key

KBTGPFZDZIZYLB-UHFFFAOYSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanhong Shan et al.
Journal of chromatography. A, 1375, 101-109 (2014-12-17)
To develop a novel hybrid monolithic column based on pentafluorobenzyl imidazolium bromide ionic liquid, a new ionic liquid monomer was synthesized from 1-vinylimidazole and pentafluorobenzyl bromide. By employing a facile one-step copolymerization of polyhedral-oligomeric-silsesquioxane-type (POSS) cross-linking agent and the home-made

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service