Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

547840

Sigma-Aldrich

Tris[N,N-bis(trimethylsilyl)amide]holmium(III)

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ho(N(Si(CH3)3)2)3
CAS Number:
Molecular Weight:
646.08
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

97%

form

solid

reaction suitability

reagent type: catalyst
core: holmium

mp

165-169 °C (lit.)

SMILES string

C[Si](C)(C)N([Ho](N([Si](C)(C)C)[Si](C)(C)C)N([Si](C)(C)C)[Si](C)(C)C)[Si](C)(C)C

InChI

1S/3C6H18NSi2.Ho/c3*1-8(2,3)7-9(4,5)6;/h3*1-6H3;/q3*-1;+3

InChI key

OKANGQPIHFPJSI-UHFFFAOYSA-N

pictograms

FlameCorrosion

signalword

Danger

Hazard Classifications

Flam. Sol. 1 - Skin Corr. 1B - Water-react 2

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

36.0 °F - closed cup

flash_point_c

2.2 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Werner Mormann et al.
Macromolecular bioscience, 9(4), 369-375 (2008-11-26)
Trimethylsilylation of cellulose in different 1,3-dialkylimidazolium ionic liquids (IL) with hexamethyldisilazane (HMDS) as a silylating agent was investigated. Trimethylsilyl (TMSi) cellulose with a degree of substitution (DS) greater than 1 is insoluble in the IL. The maximum DS obtained depends
Haoyu Tang et al.
Biomacromolecules, 11(6), 1585-1592 (2010-05-15)
A series of poly(gamma-chloropropyl-L-glutamate) (PCPLG) with controlled polymer molecular weight (MW = 5-28 kg x mol(-1)) and molecular weight distribution (PDI = 1.16-1.26) have been prepared from hexamethyldisilazane (HMDS)-mediated ring-opening polymerization (ROP) of gamma-chloropropyl-L-glutamic acid based N-carboxylanhydride (CP-NCA). CD, FTIR
Xiaoguang Li et al.
Chemical communications (Cambridge, England), 47(38), 10761-10763 (2011-08-30)
A silica microsphere suspension and a silica sol are employed in a two-step dipping process for the preparation of a superhydrophobic surface. It's not only a facile way to achieve the lotus effect, but can also create a multi-functional surface
Nondestructively creating nanojunctions by combined-dynamic-mode dip-pen nanolithography.
Guanxia Shen et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 10(13), 2226-2229 (2009-07-16)
Juliana Tsz Yan Lee et al.
Scanning, 34(1), 12-25 (2012-04-26)
Common dehydration methods of cells on biomaterials for scanning electron microscopy (SEM) include air drying, hexamethyldisilazane (HMDS) or tetramethysilane (TMS) treatment and critical point drying (CPD). On the other side, freeze-drying has been widely employed in dehydrating biological samples and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service