Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

547786

Sigma-Aldrich

Tris[N,N-bis(trimethylsilyl)amide]cerium(III)

96%

Synonym(s):

1,1,1-Trimethyl-N-(trimethylsilyl)silanamine cerium(III) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce(N(Si(CH3)3)2)3
CAS Number:
Molecular Weight:
621.27
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

96%

form

solid

reaction suitability

reagent type: catalyst
core: cerium

mp

132-140 °C (lit.)

SMILES string

C[Si](C)(C)N([Ce](N([Si](C)(C)C)[Si](C)(C)C)N([Si](C)(C)C)[Si](C)(C)C)[Si](C)(C)C

InChI

1S/3C6H18NSi2.Ce/c3*1-8(2,3)7-9(4,5)6;/h3*1-6H3;/q3*-1;+3

InChI key

DPCZDYUESIKMSU-UHFFFAOYSA-N

application

Reactant for synthesis of:
  • Amidocerium(IV) oxides
  • Sterically encumbered N-heterocyclic carbene ligands
  • Lanthanide imidodiphosphinate complexes with electroluminescent properties
  • Lanthanide complexes of an oxazoline-phenoxide hybrid chelate

Reactant for:
  • One-electron oxidation to produce tetravalent cerium compounds
  • Addition-elimination reactions across the M-C bond of metal N-heterocyclic carbenes

pictograms

FlameCorrosion

signalword

Danger

Hazard Classifications

Flam. Sol. 1 - Skin Corr. 1B - Water-react 2

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

36.0 °F - closed cup

flash_point_c

2.2 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jerry Isaacson et al.
Angewandte Chemie (International ed. in English), 48(10), 1845-1848 (2009-01-29)
(-)-Dysibetaine has been synthesized in 11 steps from readily available L-malic acid (see scheme). The key step is a unique Ugi 4-center-3-component cyclization reaction, where an ester group acts as the carboxylic acid component. The use of 1,1,1,3,3,3-hexamethyldisilazane as an
A Sebok et al.
Journal of chromatography. A, 1211(1-2), 104-112 (2008-10-14)
This paper presents a derivatization, mass fragmentation study relating to the most common six cholic acids, such as cholic, lithocholic, chenodeoxycholic, ursodeoxycholic, 3-hydroxy,7-ketocholanic and dehydrocholic acids, identified and quantified as pollutants in the aquatic environment at the first time. Derivatizations
Natalie M Clark et al.
Chemical communications (Cambridge, England), (39)(39), 5835-5837 (2009-09-30)
'Conventional' (-)-sparteine adducts of lithium and sodium 1,1,1,3,3,3-hexamethyldisilazide (HMDS) were prepared and characterised, along with an unexpected and 'unconventional' hydroxyl-incorporated sodium sodiate, [(-)-sparteine x Na(mu-HMDS)Na x (-)-sparteine](+)[Na(4)(mu-HMDS)(4)(OH)](-)--the complex anion of which is the first inverse crown ether anion.
Nur Hazlin Hazrin-Chong et al.
Journal of microbiological methods, 90(2), 96-99 (2012-05-09)
The use of hexamethyldisilazane (HMDS) as a drying agent was investigated in the specimen preparation for scanning electron microscopy (SEM) imaging of bacterial surface colonization on sub-bituminous coal. The ability of microbes to biofragment, ferment and generate methane from coal
Elsa Vennat et al.
Dental materials : official publication of the Academy of Dental Materials, 25(6), 729-735 (2009-01-29)
The objectives of this study were to assess demineralized dentin porosity and quantify the different porous features distribution within the material using mercury intrusion porosimetry (MIP) technique. We compared hexamethyldisilazane (HMDS) drying and lyophilization (LYO) (freeze-drying) in sample preparation. Fifty-six

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service