Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

544620

SAFC

Phosphorus pentasulfide

98%

Synonym(s):

Phosphorus(V) sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
P2S5
CAS Number:
Molecular Weight:
222.27
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

98%

mp

280-284 °C (lit.)

density

2.09 g/mL at 25 °C (lit.)

SMILES string

S=P(=S)SP(=S)=S

InChI

1S/P2S5/c3-1(4)7-2(5)6

InChI key

HWVAJUNEPOKXLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Flam. Sol. 1 - Water-react 1

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dominik Wohlmuth et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 16(12), 2582-2593 (2015-07-21)
The development of safe and long-lasting all-solid-state batteries with high energy density requires a thorough characterization of ion dynamics in solid electrolytes. Commonly, conductivity spectroscopy is used to study ion transport; much less frequently, however, atomic-scale methods such as nuclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service