Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

544175

Sigma-Aldrich

Lead(II) salicylate

97%

Synonym(s):

Lead salicylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(OCOC6H4OH)2
CAS Number:
Molecular Weight:
481.43
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
97%
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

assay

97%

form

powder

SMILES string

Oc1ccccc1C(=O)O[PbH2]OC(=O)c2ccccc2O

InChI

1S/2C7H6O3.Pb/c2*8-6-4-2-1-3-5(6)7(9)10;/h2*1-4,8H,(H,9,10);/q;;+2/p-2

InChI key

CNVULGHYDPMIHD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service