Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

541516

Sigma-Aldrich

2-Chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10ClO2P
CAS Number:
Molecular Weight:
168.56
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

96%

refractive index

n20/D 1.476 (lit.)

bp

46-47 °C/1.6 mmHg (lit.)

density

1.215 g/mL at 25 °C (lit.)

SMILES string

CC1(C)COP(Cl)OC1

InChI

1S/C5H10ClO2P/c1-5(2)3-7-9(6)8-4-5/h3-4H2,1-2H3

InChI key

RAOZVFGTRXLBTA-UHFFFAOYSA-N

Application

2-Chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane may be used to synthesize:
  • 3,5-di-O-benzyl-2-deoxy-4-O-(5,5-dimethyl-1,3,2-dioxaphosphorin-2-yl)-D-erythro-pentose diethyl dithioacetal[1]
  • (2-chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane) tetracarbonyliron, (2-Cl-DMP)Fe(CO)4[2]
  • bis(p-5,5-dimethyl-1,3,2-dioxaphosphorinano)-diiron hexacarbonyl, [(p-DMP)Fe(CO)3]2[2]
  • tungsten (Z)-1,2-diphosphite-alkene tetracarbonyl complexes[3]
  • tungsten (E)-vinyl-phosphite pentacarbonyl complexes[3]

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

180.0 °F - closed cup

flash_point_c

82.2 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bis(?-5, 5-dimethyl-1, 3, 2-dioxaphosphorinano)-diiron hexacarbonyl: a fluxional molecule exhibiting two types of proton site exchange.
Bartish, Charles M., and Charles S. Kraihanzel.
Inorganic Chemistry, 17(3), 735-738 (1978)
Tungsten (Z)-1, 2-diphosphite-alkene and (E)-vinyl-phosphite complexes from phosphite Fischer carbene complexes.
Barluenga J, et al.
Organometallics, 16(17), 3732-3734 (1997)
An improved synthesis of benzyl 3, 5-di-O-benzyl-2-deoxy-1, 4-dithio-d-erythro-pentofuranoside, an intermediate in the synthesis of 4'-thionucleosides.
Dyson MR, et al.
Carbohydrate Research, 216, 237-248 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service