Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

539740

Sigma-Aldrich

3-O-Chloroacetyl-5-O-benzyl-1,2-O-isopropylidene-(α)-D-xylofuranose

Synonym(s):

1,2-O-Isopropylidene-5-O-benzl-α-D-xylofuranose Chloroacetyl, Chloroacetic acid 5-benzyloxymethyl-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxo-6-yl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21ClO6
CAS Number:
Molecular Weight:
356.80
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

refractive index

n20/D 1.504 (lit.)

density

1.315 g/mL at 25 °C (lit.)

SMILES string

CC1(C)O[C@H]2O[C@H](COCc3ccccc3)[C@H](OC(=O)CCl)[C@H]2O1

InChI

1S/C17H21ClO6/c1-17(2)23-15-14(22-13(19)8-18)12(21-16(15)24-17)10-20-9-11-6-4-3-5-7-11/h3-7,12,14-16H,8-10H2,1-2H3/t12-,14+,15-,16-/m1/s1

InChI key

ZPRVBHKLXAPLQI-SLBVQIDZSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service