Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

537322

Sigma-Aldrich

Novozyme 435 (immobilized on acrylic resin)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

parameter

40-60 °C optimum reaction temp.

Application

Used for chemoenzymatic synthesis of 3-hydroxypyrrolidines and 4-(silyloxy)-2-oxopyrrolidines with two stereogenic centers.[1]

comparable product


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Víctor M. Sánchez et al.
The Journal of organic chemistry, 64(5), 1464-1470 (2001-10-25)
Aminolyses of racemic and prochiral 3-hydroxyesters with racemic amines are effectively catalyzed by Candida antarctica lipase. In these processes, the simultaneous resolution of the ester and the amine, or the corresponding asymmetrization-resolution, takes place. In all cases, the high enantioselectivity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service