Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

534048

Sigma-Aldrich

Tetrahydrofuran

histological grade, ≥99%

Synonym(s):

THF, Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein/REAXYS Number:
102391
EC Number:
MDL number:
UNSPSC Code:
12190000

grade

histological grade

vapor density

2.5 (vs air)

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

assay

≥99%

autoignition temp.

610 °F

expl. lim.

1.8-11.8 %

impurities

<0.05% Water

evapn. residue

<0.001%

refractive index

n20/D 1.407 (lit.)

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

density

0.889 g/mL at 25 °C (lit.)

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

recommended


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gregory W Charville et al.
Biomaterials, 29(30), 4039-4044 (2008-07-29)
The ability of nitric oxide (NO)-releasing xerogels to reduce fibrinogen-mediated adhesion of Staphylococcus aureus, Staphylococcus epidermidis, and Escherichia coli is described. A negative correlation was observed between NO surface flux and bacterial adhesion for each species tested. For S. aureus
Gerald I Richard et al.
Applied spectroscopy, 62(5), 476-480 (2008-05-24)
The spectroscopic properties of a chiral boronic acid based resorcinarene macrocycle employed for chiral analysis were investigated. Specifically, the emission and excitation characteristics of tetraarylboronate resorcinarene macrocycle (TBRM) and its quantum yield were evaluated. The chiral selector TBRM was investigated
Stefanie Rotter et al.
Environmental science & technology, 49(16), 10048-10056 (2015-07-22)
Aquatic ecosystems are often contaminated with large numbers of chemicals, which cannot be sufficiently addressed by chemical target analyses. Effect-directed analysis (EDA) enables the identification of toxicants in complex contaminated environmental samples. This study suggests pollution-induced community tolerance (PICT) as
T S Anirudhan et al.
Journal of colloid and interface science, 456, 22-31 (2015-06-19)
A novel adsorbent, 2-mercaptobenzamide modified itaconic acid-grafted-magnetite nanocellulose composite [P(MB-IA)-g-MNCC] was synthesized for adsorbing mercury(II) [Hg(II)] ions selectively from aqueous solutions. Fourier transforms infrared spectroscopy, X-ray diffraction, scanning electron microscopy and thermogravimetric studies were performed to characterize the adsorbent. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service