Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

529885

Sigma-Aldrich

Triphenyl(phenylethynyl)lead(IV)

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3PbC≡CC6H5
CAS Number:
Molecular Weight:
539.64
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

97%

mp

54-57 °C (lit.)

SMILES string

c1ccc(cc1)C#C[Pb](c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C8H5.3C6H5.Pb/c1-2-8-6-4-3-5-7-8;3*1-2-4-6-5-3-1;/h3-7H;3*1-5H;

InChI key

MGCZPODMNMQFSU-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

239.0 °F - closed cup

flash_point_c

115 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service