529869
Hydroxyisopropylcyclopentadienylmanganese(I) tricarbonyl
96%
About This Item
assay
96%
reaction suitability
core: manganese
mp
43-46 °C (lit.)
SMILES string
[Mn].[C-]#[O+].[C-]#[O+].[C-]#[O+].CC(C)(O)[C]1[CH][CH][CH][CH]1
InChI
1S/C8H11O.3CO.Mn/c1-8(2,9)7-5-3-4-6-7;3*1-2;/h3-6,9H,1-2H3;;;;
InChI key
VFBQMBAGBVUYSY-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 2 Oral
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
230.0 °F - closed cup
flash_point_c
110 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service