Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

52924

Sigma-Aldrich

1-Ethyl-3-methylimidazolium methanesulfonate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N2O3S
CAS Number:
Molecular Weight:
206.26
MDL number:
UNSPSC Code:
12352100

assay

≥98%

impurities

≤1.0% water

SMILES string

CS([O-])(=O)=O.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.CH4O3S/c1-3-8-5-4-7(2)6-8;1-5(2,3)4/h4-6H,3H2,1-2H3;1H3,(H,2,3,4)/q+1;/p-1

InChI key

IXLWEDFOKSJYBD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

546.8 °F - closed cup

flash_point_c

286 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service