Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

527564

Sigma-Aldrich

4-Ethoxycarbonylphenylboronic acid pinacol ester

≥96%

Synonym(s):

4-Carboethoxyphenylboronic acid pinacol ester, Ethyl 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21BO4
CAS Number:
Molecular Weight:
276.14
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

≥96%

refractive index

n20/D 1.5 (lit.)

bp

330 °C (lit.)

density

1.052 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)c1ccc(cc1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C15H21BO4/c1-6-18-13(17)11-7-9-12(10-8-11)16-19-14(2,3)15(4,5)20-16/h7-10H,6H2,1-5H3

InChI key

NCVIYKCFTYSAGN-UHFFFAOYSA-N

Analysis Note

Reactant for:
  • Oxidative Heck cross-coupling reactions
  • Suzuki-Miyaura coupling

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service