Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

527300

Sigma-Aldrich

PSS-Diphenylvinylsilyloxy-heptacyclopentyl substituted

Synonym(s):

(Diphenylvinylsilyloxy)heptacyclopentylpentacyclooctasiloxane, Diphenylvinylsilyloxy-POSS®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H76O13Si9
CAS Number:
Molecular Weight:
1125.89
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

mp

211-216 °C (lit.)

SMILES string

C=C[Si](O[Si]12O[Si]3(O[Si]4(O[Si](O1)(O[Si]5(O[Si](O2)(O[Si](O3)(O[Si](O4)(O5)C6CCCC6)C7CCCC7)C8CCCC8)C9CCCC9)C%10CCCC%10)C%11CCCC%11)C%12CCCC%12)(c%13ccccc%13)c%14ccccc%14

InChI

1S/C49H76O13Si9/c1-2-63(41-23-5-3-6-24-41,42-25-7-4-8-26-42)50-71-60-68(47-35-17-18-36-47)54-65(44-29-11-12-30-44)51-64(43-27-9-10-28-43)52-66(56-68,45-31-13-14-32-45)58-70(62-71,49-39-21-22-40-49)59-67(53-64,46-33-15-16-34-46)57-69(55-65,61-71)48-37-19-20-38-48/h2-8,23-26,43-49H,1,9-22,27-40H2

InChI key

RPKVGQZTLKOUOW-UHFFFAOYSA-N

Legal Information

POSS is a registered trademark of Hybrid Plastics, Inc.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service