525820
1-Propanethiol
Synonym(s):
NPM™, n-Propylmercaptan, Mercaptan C3
About This Item
Recommended Products
vapor density
2.54 (vs air)
vapor pressure
122 mmHg ( 20 °C)
availability
available only in EU
refractive index
n20/D 1.426 (lit.)
bp
67-68 °C (lit.)
mp
−113 °C (lit.)
density
0.820 g/mL at 25 °C (lit.)
SMILES string
CCCS
InChI
1S/C3H8S/c1-2-3-4/h4H,2-3H2,1H3
InChI key
SUVIGLJNEAMWEG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2 - Skin Sens. 1B
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
-4.0 °F - closed cup
flash_point_c
-20 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service