Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

525782

Sigma-Aldrich

2-Ethylhexyl thioglycolate

Synonym(s):

2-EHTG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2COOCH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
204.33
Beilstein/REAXYS Number:
9047474
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

bp

255-260 °C (lit.)

density

0.972 g/mL at 20 °C (lit.)

SMILES string

CCCCC(CC)COC(=O)CS

InChI

1S/C10H20O2S/c1-3-5-6-9(4-2)7-12-10(11)8-13/h9,13H,3-8H2,1-2H3

InChI key

OWHSTLLOZWTNTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of Arkema

replaced by

Product No.
Description
Pricing

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

243.5 °F - Pensky-Martens closed cup

flash_point_c

117.5 °C - Pensky-Martens closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neepa Choksi et al.
Cutaneous and ocular toxicology, 39(3), 180-192 (2020-06-27)
OptiSafe is an in chemico test method that identifies potential eye irritants based on macromolecular damage following test chemical exposure. The OptiSafe protocol includes a prescreen assessment that identifies test chemicals that are outside the applicability domain of the test

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service