Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

52381

Sigma-Aldrich

Hexadecyltrimethylammonium hydroxide hydrate

≥98.0% (calc. based on dry substance, T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15N(OH)(CH3)3 · xH2O
CAS Number:
Molecular Weight:
301.55 (anhydrous basis)
Beilstein/REAXYS Number:
3723055
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

assay

≥98.0% (calc. based on dry substance, T)

impurities

≤0.1% halogenides (as chloride)
~15% water

storage temp.

2-8°C

SMILES string

O.[OH-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Looking for similar products? Visit Product Comparison Guide

Other Notes

Crystalline quaternary ammonium hydroxide; CMC: 1 mM, aggregation number: 46-51[1]

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ingvarsson, A., et al.
Analytical Chemistry, 60, 2047-2047 (1988)
Yoshinori Iiguni et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 29(1), 35-39 (2013-01-11)
By using the electromagnetophoretic migration technique with permanent magnets, the electromagnetophoretic migration velocimetry in a droplet-based system was demonstrated for organic droplets dispersed in an aqueous solution. Migration of 2-fluorotoluene droplets with a diameter of 8 - 16 μm dispersed
Gurinder Jit Randhawa et al.
Methods in molecular biology (Clifton, N.J.), 958, 139-151 (2012-11-13)
The adoption rate and global area under cultivation of genetically modified (GM) crops is dramatically increasing in recent past. GM cotton has occupied 25.0 million hectares (mha) comprising 15.6% of the global area under GM cultivation. Bt cotton, expressing delta-endotoxins
Tanja Mehling et al.
Journal of chromatography. A, 1273, 66-72 (2013-01-01)
Several methods for the description of the retention behavior in micellar liquid chromatography (MLC) were described previously. Thereby, the most common are the linear solvation energy relationships (LSER). However, for the evaluation of the LSER, a number of experimental data
Ahmet Gürses et al.
TheScientificWorldJournal, 2012, 270452-270452 (2013-02-01)
The aim of this study was the preparation of polyethylene oxide (PEO)/clay nanocomposites using organoclay produced via micellar adsorption of cethyltrimethyl ammonium bromide (CTAB) and their characterisation by X-ray diffraction (XRD), and Fourier transform infrared (FT-IR) spectra, and the investigation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service