Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

522287

Sigma-Aldrich

Tributylmethylammonium hydroxide solution

20 wt. % in H2O

Synonym(s):

Methyltributylammonium hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3N(OH)CH3
CAS Number:
Molecular Weight:
217.39
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

concentration

20 wt. % in H2O

refractive index

n20/D 1.374

bp

102 °C

density

0.993 g/mL at 25 °C

SMILES string

[OH-].CCCC[N+](C)(CCCC)CCCC

InChI

1S/C13H30N.H2O/c1-5-8-11-14(4,12-9-6-2)13-10-7-3;/h5-13H2,1-4H3;1H2/q+1;/p-1

InChI key

QVOFCQBZXGLNAA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

132.8 °F - closed cup

flash_point_c

56 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J W Smit et al.
The Journal of pharmacology and experimental therapeutics, 286(1), 321-327 (1998-07-10)
We recently showed that absence of mdr1-type P-glycoprotein (P-gp) in mice resulted in profoundly reduced hepatic and intestinal clearance of type 1 and type 2 cationic drugs compared with that in wild-type mice. These data strongly support the concept that
H Steen et al.
Biochemical pharmacology, 44(12), 2323-2331 (1992-12-15)
To test the hypothesis of multiplicity in carrier-mediated uptake mechanisms for organic cations in the liver and to study the possible relation with bile acid and cardiac glycoside uptake mechanisms, mutual interaction during uptake of various radiolabeled quaternary amines has
Soon-Sun Hong et al.
Archives of pharmacal research, 29(4), 318-322 (2006-05-10)
Many quaternary ammonium salts are incompletely absorbed after their oral administration and may also be actively secreted into the intestine. However, the underlying mechanism(s) that control the transport of these cations across the intestinal epithelium is not well understood. In
Soon-Sun Hong et al.
Archives of pharmacal research, 29(4), 323-327 (2006-05-10)
The objective of this study was to examine the pharmacokinetics of organic cations in intrahepatic cholestatic rats. A pretreatment with 17alpha-ethynylestradiol was used to induce intrahepatic cholestasis, and tributylmethylammonium (TBuMA) was used as a representative model organic cation. When [3H]TBuMA
Min-Koo Choi et al.
Journal of pharmaceutical sciences, 94(2), 317-326 (2004-12-01)
The in vivo canalicular excretion clearance of tributylmethyl ammonium (TBuMA), a P-glycoprotein (P-gp) substrate, was previously reported to be unaffected by the induction of an experimental hepatic injury (EHI) by CCl(4) despite the increased expression of P-gp in the EHI

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service