Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

517836

Sigma-Aldrich

Tris(diethylamido)(ethylimido)tantalum(V)

99.99% trace metals basis

Synonym(s):

Tris(diethylamino)(ethylimino)tantalum(V)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5NTa(N(C2H5)2)3
CAS Number:
Molecular Weight:
440.40
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

99.99% trace metals basis

reaction suitability

core: tantalum
reagent type: catalyst

refractive index

n20/D 1.51 (lit.)

density

1.31 g/mL at 25 °C (lit.)

SMILES string

CCN=[Ta](N(CC)CC)(N(CC)CC)N(CC)CC

InChI

1S/3C4H10N.C2H5N.Ta/c3*1-3-5-4-2;1-2-3;/h3*3-4H2,1-2H3;2H2,1H3;/q3*-1;;+3

InChI key

ZLKUSFBEBZOVGX-UHFFFAOYSA-N

General description

Atomic number of base material: 73 Tantalum

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

36.0 °F - closed cup

flash_point_c

2.2 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service