Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

517801

Sigma-Aldrich

2,4,6,8,10-Pentamethylcyclopentasiloxane

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3SiHO)5
CAS Number:
Molecular Weight:
300.64
Beilstein/REAXYS Number:
1792134
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

96%

form

liquid

refractive index

n20/D 1.392 (lit.)

bp

54 °C/10 mmHg (lit.)

density

0.992 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[SiH]1O[SiH](C)O[SiH](C)O[SiH](C)O[SiH](C)O1

InChI

1S/C5H20O5Si5/c1-11-6-12(2)8-14(4)10-15(5)9-13(3)7-11/h11-15H,1-5H3

InChI key

HGPDWMTUQRNTQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

98.6 °F - closed cup

flash_point_c

37 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

atomic layer deposition (ALD), microelectronics, Mo:Al2O3 films, nanocomposite coating, photovoltaics, semiconductor devices, W:Al2O3 films, composite films, layer-by-layer

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Synthesis of Melting Gels Using Mono-Substituted and Di-Substituted Alkoxysiloxanes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service