516856
Thioglycolic acid
80%
Synonym(s):
Mercaptoacetic acid
About This Item
Recommended Products
vapor density
3.2 (vs air)
vapor pressure
0.4 mmHg ( 25 °C)
assay
80%
autoignition temp.
662 °F
refractive index
n20/D 1.505 (lit.)
bp
96 °C/5 mmHg (lit.)
mp
−16 °C (lit.)
density
1.326 g/mL at 20 °C (lit.)
SMILES string
OC(=O)CS
InChI
1S/C2H4O2S/c3-2(4)1-5/h5H,1H2,(H,3,4)
InChI key
CWERGRDVMFNCDR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1B
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 1
flash_point_f
266.0 °F - closed cup
flash_point_c
130 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service