Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

516341

Sigma-Aldrich

(R)-(+)-N-Boc-2-piperidinecarboxylic acid

98%

Synonym(s):

Boc-D-Pip-OH, (+)-N-Boc-(R)-pipecolinic acid, (R)-(+)-1-(tert-Butoxycarbonyl)-2-piperidinecarboxylic acid, (R)-N-Boc-piperidine-2-carboxylic acid, N-Boc-D-pipecolic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO4
CAS Number:
Molecular Weight:
229.27
Beilstein/REAXYS Number:
480023
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

assay

98%

optical activity

[α]23/D 63.5±3°, c = 1 in acetic acid

optical purity

ee: 98% (GLC)

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

116-119 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N1CCCC[C@@H]1C(O)=O

InChI

1S/C11H19NO4/c1-11(2,3)16-10(15)12-7-5-4-6-8(12)9(13)14/h8H,4-7H2,1-3H3,(H,13,14)/t8-/m1/s1

InChI key

JQAOHGMPAAWWQO-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service