515957
Merrifield′s peptide resin
100-200 mesh, extent of labeling: 1.0-1.5 mmol/g Cl- loading, 1 % cross-linked
Synonym(s):
Chloromethylated polystyrene, Chloromethylated stryene/divinylbenzene copolymer, Chloromethylated styrene-divinylbenzene resin, Copolymer of styrene and divinylbenzene, chloromethylated, Poly(styrene-co-vinylbenzyl chloride-co-divinylbenzene), Polystyrene crosslinked with divinylbenzene, chloromethylated
About This Item
crosslinking
1 % cross-linked
extent of labeling
1.0-1.5 mmol/g Cl- loading
particle size
100-200 mesh
SMILES string
ClCc1ccccc1.C=Cc2ccccc2.C=Cc3ccc(C=C)cc3
InChI
1S/CH3Cl/c1-2/h1H3
InChI key
NEHMKBQYUWJMIP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service