515787
α,β-Bis(phenylazo)stilbene, mixture of isomers
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5N=NC(C6H5)=C(C6H5)N=NC6H5
CAS Number:
Molecular Weight:
388.46
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
impurities
<10% benzil bis(phenylhydrazone)
mp
162 °C (dec.) (lit.)
SMILES string
c1ccc(cc1)\N=N\C(c2ccccc2)=C(\N=N\c3ccccc3)c4ccccc4
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service