Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

514063

Sigma-Aldrich

Ytterbium(III) isopropoxide

99.9% trace metals basis

Synonym(s):

Tris(tetramethylcyclopentadienyl)lanthanum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHO]3Yb
CAS Number:
Molecular Weight:
350.30
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

assay

99.9% trace metals basis

form

solid

reaction suitability

core: ytterbium
reagent type: catalyst

mp

349 °C (dec.) (lit.)

SMILES string

CCCO[Yb](OCCC)OCCC

InChI

1S/3C3H7O.Yb/c3*1-2-3-4;/h3*2-3H2,1H3;/q3*-1;+3

InChI key

VKLVFRMIYPCEHG-UHFFFAOYSA-N

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Sol. 1 - Skin Corr. 1B

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carmen Cavallo et al.
Scientific reports, 5, 16785-16785 (2015-11-19)
Solid solutions of the rare earth (RE) cations Pr(3+), Nd(3+), Sm(3+), Gd(3+), Er(3+) and Yb(3+) in anatase TiO2 have been synthesized as mesoporous beads in the concentration range 0.1-0.3% of metal atoms. The solid solutions were have been characterized by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service