Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

513520

Sigma-Aldrich

Luperox® 101XL45, 2,5-Bis(tert-butylperoxy)-2,5-dimethylhexane, blend with calcium carbonate and silica

Synonym(s):

2,5-Bis(tert-butylperoxy)-2,5-dimethylhexane, blend with calcium carbonate and silica

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3COOC(CH3)2CH2-]2
CAS Number:
Molecular Weight:
290.44
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

concentration

~45 wt. %

SMILES string

CC(C)(C)OOC(C)(C)CCC(C)(C)OOC(C)(C)C

InChI

1S/C16H34O4/c1-13(2,3)17-19-15(7,8)11-12-16(9,10)20-18-14(4,5)6/h11-12H2,1-10H3

InChI key

DMWVYCCGCQPJEA-UHFFFAOYSA-N

Application

Polymerization initiator

Legal Information

Product of Arkema Inc.
Luperox is a registered trademark of Arkema Inc.

signalword

Warning

Hazard Classifications

Muta. 2 - Org. Perox. E - Skin Irrit. 2

Storage Class

5.2 - Organic peroxides and self-reacting hazardous materials

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service