513296
Cumene hydroperoxide (Luperox® CU90)
88%
Synonym(s):
Cumene hydroperoxide, α,α-Dimethylbenzyl hydroperoxide
About This Item
Recommended Products
vapor density
5.4 (vs air)
vapor pressure
<0.03 mmHg ( 20 °C)
assay
88%
reaction suitability
reagent type: oxidant
refractive index
n20/D 1.5230
bp
100-101 °C/8 mmHg (lit.)
101 °C/8 mmHg
density
1.038 g/mL at 25 °C
SMILES string
CC(C)(OO)c1ccccc1
InChI
1S/C9H12O2/c1-9(2,11-10)8-6-4-3-5-7-8/h3-7,10H,1-2H3
InChI key
YQHLDYVWEZKEOX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
signalword
Danger
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Org. Perox. F - Skin Corr. 1B - STOT RE 2 Inhalation - STOT SE 3
target_organs
Lungs, Respiratory system
Storage Class
5.2 - Organic peroxides and self-reacting hazardous materials
wgk_germany
WGK 3
flash_point_f
161.6 °F
flash_point_c
72 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service