Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51241

Sigma-Aldrich

Ethyl (S)-2-oxiranylacetate

≥96.0% (sum of enantiomers, GC)

Synonym(s):

Ethyl (S)-3,4-epoxybutyrate, Ethyl (S)-oxirane-2-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O3
CAS Number:
Molecular Weight:
130.14
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

assay

≥96.0% (sum of enantiomers, GC)

optical purity

enantiomeric ratio: ≥99:1

storage temp.

2-8°C

SMILES string

CCOC(=O)C[C@H]1CO1

InChI

1S/C6H10O3/c1-2-8-6(7)3-5-4-9-5/h5H,2-4H2,1H3/t5-/m0/s1

InChI key

WHUSTVAXKRFVPD-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service