Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

511374

Sigma-Aldrich

Tris(tetramethylcyclopentadienyl)europium(III)

99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H39Eu
CAS Number:
Molecular Weight:
515.56
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

assay

99.9%

reaction suitability

core: europium
reagent type: catalyst

SMILES string

[Eu].C[C]1[CH][C](C)[C](C)[C]1C.C[C]2[CH][C](C)[C](C)[C]2C.C[C]3[CH][C](C)[C](C)[C]3C

InChI

1S/3C9H13.Eu/c3*1-6-5-7(2)9(4)8(6)3;/h3*5H,1-4H3;

InChI key

SEHXYMOUURJNBC-UHFFFAOYSA-N

General description

Atomic number of base material: 63 Europium

Application

Reactant for synthesis of a mini-fullerene type zintl anion containing a lanthanide ion

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Sol. 1

supp_hazards

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

High Purity Metalorganic Precursors for CPV Device Fabrication

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service