Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

511048

Sigma-Aldrich

Allyl(4-methoxyphenyl)dimethylsilane

98%

Synonym(s):

Allyldimethyl(4-methoxyphenyl)silane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4Si(CH3)2CH2CH=CH2
CAS Number:
Molecular Weight:
206.36
Beilstein/REAXYS Number:
2830799
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

98%

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.519 (lit.)

bp

253 °C (lit.)

density

0.935 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(cc1)[Si](C)(C)CC=C

InChI

1S/C12H18OSi/c1-5-10-14(3,4)12-8-6-11(13-2)7-9-12/h5-9H,1,10H2,2-4H3

InChI key

BEKRXBQZYQZFBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

215.1 °F - closed cup

flash_point_c

101.7 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service